Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002834.3 | NP_002825.3 | 16 | 5418 | ||
XM_006719526.2 | XP_006719589.1 | 16 | 5208 | |||
XM_011538613.2 | XP_011536915.1 | 16 | 5244 | |||
XM_017019722.1 | XP_016875211.1 | 16 | 5235 | |||
GenBank mRNA | BC007869.2 | - | 1 | 907 | ||
BC025181.1 | - | 1 | 983 | |||
BC030949.1 | - | 1 | 928 | |||
BX537632.1 | - | 1 | 2507 | |||
U79291.1 | - | 1 | 478 |