Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBL1Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033284.1 | NP_150600.1 | 7 | 838 | ||
NM_134258.1 | NP_599020.1 | 6 | 807 | |||
NM_134259.1 | NP_599021.1 | 6 | 743 | |||
XM_005262572.3 | XP_005262629.1 | 1 | 502 | |||
XM_017030086.1 | XP_016885575.1 | 7 | 827 | |||
XM_017030087.1 | XP_016885576.1 | 7 | 827 | |||
GenBank mRNA | AF332220.1 | - | 7 | 838 | ||
AF332221.1 | - | 6 | 807 | |||
AF332222.1 | - | 6 | 743 | |||
AK298210.1 | - | 6 | 719 | |||
BC130471.1 | - | 3 | 321 | 40146502 | ||
BC130473.1 | - | 3 | 321 | 40146504 |