Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303511.1 | NP_001290440.1 | 3 | 342 | ||
NM_005230.3 | NP_005221.2 | 5 | 1579 | |||
XM_006719275.3 | XP_006719338.1 | 6 | 1676 | |||
GenBank mRNA | AL705041.1 | - | 3 | 342 | ||
BC017371.1 | - | 5 | 1461 | 4287696 | ||
Z36715.1 | - | 5 | 1532 |