Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DACH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139514.1 | NP_001132986.1 | 1 | 409 | ||
NM_053281.3 | NP_444511.1 | 1 | 409 | |||
XM_011530846.2 | XP_011529148.1 | 1 | 408 | |||
XM_011530847.2 | XP_011529149.1 | 1 | 408 | |||
XM_011530848.2 | XP_011529150.1 | 1 | 408 | |||
XM_017029254.1 | XP_016884743.1 | 1 | 408 | |||
XM_017029255.1 | XP_016884744.1 | 1 | 408 | |||
XM_017029256.1 | XP_016884745.1 | 1 | 408 | |||
GenBank mRNA | AF428101.1 | - | 1 | 239 | ||
AK091842.1 | - | 1 | 402 | |||
AK290596.1 | - | 1 | 409 | |||
BC114950.1 | - | 1 | 239 | 40006598 |