Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC36A1 SIRNA Assays ›
Gene Symbol:
SLC36A1
Gene Name:
solute carrier family 36 member 1
Gene Aliases:
Dct1, LYAAT1, PAT1, TRAMD3
Chromosome Location:
Chr.5: 151411344 - 151556318 on Build GRCh38
UniGene ID:
Hs.269004
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
35905
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC36A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308150.1
NP_001295079.1
2
313
NM_001308151.1
NP_001295080.1
2
283
NM_078483.3
NP_510968.2
2
313
XM_005268386.1
XP_005268443.1
2
283
XM_006714759.3
XP_006714822.1
2
316
XM_011537580.2
XP_011535882.1
2
1003
XM_011537581.1
XP_011535883.1
3
431
XM_011537583.2
XP_011535885.1
3
4868
XM_011537584.2
XP_011535886.1
2
522
XM_011537585.1
XP_011535887.1
3
378
XM_011537586.2
XP_011535888.1
2
1126
XM_011537587.2
XP_011535889.1
2
382
XM_011537588.2
XP_011535890.1
2
226
XM_011537589.2
XP_011535891.1
2
1134
XM_011537590.1
XP_011535892.1
2
141
XM_011537591.1
XP_011535893.1
2
116
XM_011537592.2
XP_011535894.1
2
120
XM_011537593.2
XP_011535895.1
2
317
XM_011537594.1
XP_011535896.1
3
431
XM_011537595.2
XP_011535897.2
2
1009
XM_011537596.2
XP_011535898.2
1
95
XM_017009216.1
XP_016864705.1
2
209
XM_017009217.1
XP_016864706.1
2
313
XM_017009218.1
XP_016864707.1
1
95
XM_017009219.1
XP_016864708.1
2
317
XM_017009220.1
XP_016864709.1
2
283
GenBank mRNA
AF516142.1
-
1
105
AK057340.1
-
2
283
AK309323.1
-
2
285
AY162213.1
-
2
218
AY227111.1
-
3
233
AY227113.1
-
2
189
BC136437.1
-
2
184
9020426
BC136438.1
-
2
184
9020427
BM787446.1
-
2
316
BX537963.1
-
2
310
GU296670.1
-
1
90
Back To Top
More Information
Panther Classification:
Molecular Function -
amino acid transporter
Gene Ontology Categories:
Function(s)
Process(es)
ion transport
amino acid transport
L-alanine transport
glycine transport
proline transport
proline transmembrane transport
hydrogen ion transmembrane transport
hydrogen:amino acid symporter activity
hydrogen ion transmembrane transporter activity
amino acid transmembrane transporter activity
L-alanine transmembrane transporter activity
glycine transmembrane transporter activity
L-proline transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0