Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEU4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167599.2 | NP_001161071.1 | 4 | 1875 | ||
NM_001167600.2 | NP_001161072.1 | 4 | 1552 | |||
NM_001167601.2 | NP_001161073.1 | 4 | 1555 | |||
NM_001167602.2 | NP_001161074.1 | 5 | 2053 | |||
NM_080741.3 | NP_542779.2 | 4 | 1872 | |||
GenBank mRNA | AJ277883.1 | - | 3 | 1339 | ||
AK025617.1 | - | 5 | 1805 | |||
AK091038.1 | - | 5 | 2050 | |||
AK096992.1 | - | 2 | 2665 | |||
AK289421.1 | - | 4 | 1429 | |||
AK290076.1 | - | 4 | 1412 | |||
AK298403.1 | - | 5 | 1384 | |||
AK300969.1 | - | 4 | 1425 | |||
AY203952.1 | - | 4 | 1407 | |||
BC012899.1 | - | 4 | 1826 | 4156395 | ||
BC105974.1 | - | 4 | 1388 | 4944243 | ||
BC117214.1 | - | 4 | 1516 | 40125765 |