Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TJAP1 SIRNA Assays ›
          Gene Symbol:
          TJAP1
          Gene Name:
          tight junction associated protein 1
          Gene Aliases:
          PILT, TJP4
          Chromosome Location:
          Chr.6: 43477523 - 43506557 on Build GRCh38
          UniGene ID:
          Hs.520145
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 36027

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TJAP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001146016.1 NP_001139488.1 6 522
          NM_001146017.1 NP_001139489.1 5 487
          NM_001146018.1 NP_001139490.1 7 579
          NM_001146019.1 NP_001139491.1 6 522
          NM_001146020.1 NP_001139492.1 5 373
          NM_080604.2 NP_542171.2 6 574
          NR_027394.1 5 409
          XM_006715249.1 XP_006715312.1 6 500
          XM_006715250.3 XP_006715313.1 7 519
          XM_006715251.2 XP_006715314.1 6 527
          XM_006715252.2 XP_006715315.1 7 587
          XM_006715254.3 XP_006715317.1 6 425
          XM_006715256.1 XP_006715319.1 6 422
          XM_006715257.2 XP_006715320.1 6 476
          XM_006715259.3 XP_006715322.1 5 326
          XM_006715260.1 XP_006715323.1 6 443
          XM_006715261.1 XP_006715324.1 5 347
          XM_006715262.2 XP_006715325.1 6 672
          XM_006715263.2 XP_006715326.1 5 382
          XM_006715265.1 XP_006715328.1 6 422
          XM_006715266.1 XP_006715329.1 5 330
          XM_006715267.2 XP_006715330.1 6 511
          XM_006715268.2 XP_006715331.1 6 508
          XM_006715269.2 XP_006715332.1 6 563
          XM_006715270.1 XP_006715333.1 6 511
          XM_011514995.1 XP_011513297.1 7 562
          XM_011514996.1 XP_011513298.1 6 482
          XM_017011488.1 XP_016866977.1 5 400
          XM_017011489.1 XP_016866978.1 6 697
          XM_017011490.1 XP_016866979.1 6 500
          XM_017011491.1 XP_016866980.1 7 536
          XM_017011492.1 XP_016866981.1 8 665
          XM_017011493.1 XP_016866982.1 8 663
          GenBank mRNA AK024269.1 - 6 524
          AK123398.1 - 4 1676
          AK127338.1 - 5 822
          AK225787.1 - 6 475
          AK307483.1 - 5 316
          AK308866.1 - 5 404
          AK309119.1 - 5 362
          BC046239.1 - 6 442 4906152
          BC064401.1 - 6 453 5539930
          CR749480.1 - 3 1913
          DA018836.1 - 7 582
          DA051961.1 - 5 326
          DA297698.1 - 5 294

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          tight junction

          Gene Ontology Categories:

          Function(s) Process(es)

          Golgi organization
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0