Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC46A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242366.2 | NP_001229295.1 | 2 | 897 | ||
NM_080669.5 | NP_542400.2 | 2 | 897 | |||
XM_005277786.3 | XP_005277843.1 | 2 | 897 | |||
XM_017024110.1 | XP_016879599.1 | 3 | 856 | |||
GenBank mRNA | AK054669.1 | - | 2 | 895 | ||
AK097194.1 | - | 1 | 38 | |||
AK295883.1 | - | 2 | 853 | |||
AL832613.2 | - | 1 | 205 | |||
BC010691.1 | - | 2 | 886 | 3872267 |