Search
Search

CFAP36| Gene Symbol: CFAP36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282761.1 | NP_001269690.1 | 7, 8 | 817 | ||
| NM_080667.6 | NP_542398.3 | 6, 7 | 742 | |||
| XM_011532499.1 | XP_011530801.1 | 3, 4 | 999 | |||
| XM_017003254.1 | XP_016858743.1 | 5, 6 | 618 | |||
| XM_017003255.1 | XP_016858744.1 | 5, 6 | 641 | |||
| GenBank mRNA | AK309764.1 | - | 6, 7 | 691 | ||
| AY358097.1 | - | 7, 8 | 809 | |||
| BC001797.1 | - | 672 | ||||
| BC010011.2 | - | 6, 7 | 649 | 4309613 | ||