Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSC4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080744.1 | NP_542782.1 | 5 | 859 | ||
XM_017011750.1 | XP_016867239.1 | 2 | 648 | |||
GenBank mRNA | AJ312755.1 | - | 5 | 859 | ||
AK172783.1 | - | 5 | 817 | |||
BC113840.1 | - | 5 | 742 |