Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080860.3 | NP_543136.1 | 2 | 312 | ||
XM_011529786.1 | XP_011528088.1 | 2 | 204 | |||
GenBank mRNA | AB006536.1 | - | 2 | 312 | ||
AB041016.1 | - | 2 | 249 | |||
AK314746.1 | - | 2 | 149 | |||
BC101519.1 | - | 2 | 191 | 8069025 | ||
BC113367.1 | - | 2 | 191 | 8322419 |