siRNA Details
Gene Symbol: SOX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000346.3 | NP_000337.1 | 2 | 881 | ||
GenBank mRNA | AK295455.1 | - | 2 | 878 | ||
BC007951.2 | - | 2 | 738 | 4299305 | ||
BC056420.1 | - | 2 | 891 | 6200521 | ||
BT006875.1 | - | 2 | 509 | |||
Z46629.1 | - | 2 | 868 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
