Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146726.1 | NP_001140198.1 | 7, 8 | 1033 | ||
NM_138379.2 | NP_612388.2 | 8, 9 | 1117 | |||
XM_017010021.1 | XP_016865510.1 | 6, 7 | 923 | |||
GenBank mRNA | AK131025.1 | - | 4, 5 | 375 | ||
AK301337.1 | - | 7, 8 | 1004 | |||
AK312746.1 | - | 8, 9 | 1087 | |||
BC008988.1 | - | 8, 9 | 1117 | 4184237 | ||
JX049980.1 | - | 8, 9 | 1060 | |||
KU178832.1 | - | 8, 9 | 1060 | |||
KU178833.1 | - | 7, 8 | 979 |