Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SGO1 SIRNA Assays ›
Gene Symbol:
SGO1
Gene Name:
shugoshin 1
Gene Aliases:
CAID, NY-BR-85, SGO, SGOL1
Chromosome Location:
Chr.3: 20160593 - 20188143 on Build GRCh38
UniGene ID:
Hs.105153
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
36832
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SGO1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001012409.3
NP_001012409.1
2
300
NM_001012410.4
NP_001012410.1
2
300
NM_001012411.3
NP_001012411.1
2
300
NM_001012412.4
NP_001012412.1
2
300
NM_001012413.3
NP_001012413.1
2
300
NM_001199251.2
NP_001186180.1
2
411
NM_001199252.2
NP_001186181.1
2
411
NM_001199253.2
NP_001186182.1
2
411
NM_001199254.2
NP_001186183.1
2
411
NM_001199255.2
NP_001186184.1
2
411
NM_001199256.2
NP_001186185.1
2
411
NM_001199257.2
NP_001186186.1
2
411
NM_138484.4
NP_612493.1
2
300
NR_131179.1
2
300
NR_131180.1
2
300
XM_011533373.2
XP_011531675.1
2
384
XM_011533375.2
XP_011531677.1
2
753
XM_011533376.2
XP_011531678.1
2
608
XM_011533377.2
XP_011531679.1
2
1588
GenBank mRNA
AB187577.1
-
2
258
AB187578.1
-
2
258
AB187579.1
-
2
258
AB187580.1
-
2
258
AB187581.1
-
2
258
AB187582.1
-
2
258
AB190994.1
-
2
148
AB193056.1
-
2
305
AB193057.1
-
2
194
AB193058.1
-
2
194
AB193059.1
-
2
305
AB193060.1
-
2
305
AB193061.1
-
2
194
AB193062.1
-
2
305
AB193063.1
-
2
194
AB193064.1
-
2
305
AB193065.1
-
2
305
AB193066.1
-
2
194
AB451340.1
-
1
118
AB451489.1
-
1
118
AB567656.1
-
2
258
AB567657.1
-
2
258
AK308095.1
-
2
399
BC001339.2
-
1
59
BC017867.1
-
2
258
4685140
BG530826.1
-
2
272
BX494477.1
-
2
414
BX648516.1
-
2
385
DB163747.1
-
2
410
Back To Top
More Information
Panther Classification:
Molecular Function -
centromere DNA-binding protein
Gene Ontology Categories:
Function(s)
Process(es)
chromosome segregation
sister chromatid cohesion
mitotic nuclear division
attachment of spindle microtubules to kinetochore
centriole-centriole cohesion
meiotic chromosome segregation
cell division
protein binding
kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8xc7s:80/100.66.76.248:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0