Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TACC2 SIRNA Assays ›
          Gene Symbol:
          TACC2
          Gene Name:
          transforming acidic coiled-coil containing protein 2
          Gene Aliases:
          AZU-1, ECTACC
          Chromosome Location:
          Chr.10: 121989174 - 122254545 on Build GRCh38
          UniGene ID:
          Hs.501252
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 3684

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TACC2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291876.1 NP_001278805.1 7, 8 6204
          NM_001291877.1 NP_001278806.1 7, 8 6351
          NM_001291878.1 NP_001278807.1 2, 3 712
          NM_001291879.1 NP_001278808.1 4, 5 562
          NM_006997.3 NP_008928.1 2, 3 712
          NM_206860.2 NP_996742.1 2, 3 712
          NM_206861.2 NP_996743.1 5, 6 777
          NM_206862.3 NP_996744.3 8, 9 6339
          XM_005269388.1 XP_005269445.1 9, 10 6163
          XM_005269389.4 XP_005269446.1 9, 10 6687
          XM_005269390.1 XP_005269447.1 9, 10 6163
          XM_005269391.1 XP_005269448.1 8, 9 6103
          XM_005269392.1 XP_005269449.1 9, 10 6163
          XM_005269393.1 XP_005269450.1 9, 10 6163
          XM_005269394.1 XP_005269451.1 8, 9 6037
          XM_005269395.1 XP_005269452.1 8, 9 6028
          XM_005269396.1 XP_005269453.1 9, 10 6163
          XM_005269397.1 XP_005269454.1 7, 8 5902
          XM_005269399.1 XP_005269456.1 8, 9 6028
          XM_006717548.1 XP_006717611.1 9, 10 6163
          XM_006717552.1 XP_006717615.1 9, 10 6163
          XM_006717553.1 XP_006717616.1 9, 10 6163
          XM_006717554.1 XP_006717617.1 9, 10 6163
          XM_011539126.1 XP_011537428.1 9, 10 6160
          XM_011539128.1 XP_011537430.1 7, 8 736
          XM_011539129.1 XP_011537431.1 6, 7 601
          XM_011539130.1 XP_011537432.1 6, 7 601
          XM_011539131.1 XP_011537433.1 2, 3 574
          XM_011539132.1 XP_011537434.1 2, 3 574
          XM_011539133.1 XP_011537435.1 3, 4 515
          XM_011539134.1 XP_011537436.1 3, 4 786
          XM_017015484.1 XP_016870973.1 9, 10 6163
          XM_017015485.1 XP_016870974.1 9, 10 6163
          XM_017015486.1 XP_016870975.1 9, 10 6163
          XM_017015487.1 XP_016870976.1 9, 10 6163
          XM_017015488.1 XP_016870977.1 7, 8 5902
          XM_017015489.1 XP_016870978.1 7, 8 5902
          XM_017015491.1 XP_016870980.1 2, 3 714
          XM_017015492.1 XP_016870981.1 2, 3 714
          XM_017015493.1 XP_016870982.1 2, 3 574
          XM_017015494.1 XP_016870983.1 2, 3 714
          XM_017015495.1 XP_016870984.1 2, 3 574
          XM_017015496.1 XP_016870985.1 3, 4 515
          XM_017015497.1 XP_016870986.1 3, 4 786
          XM_017015498.1 XP_016870987.1 2, 3 651
          XM_017015499.1 XP_016870988.1 4, 5 755
          GenBank mRNA AF095791.2 - 2, 3 299
          AF176646.1 - 3, 4 700
          AF220152.2 - 1 376
          AF528098.1 - 5, 6 758
          AF528099.1 - 8, 9 6320
          AK295631.1 - 2, 3 495
          AL833304.1 - 7, 8 5438
          BC039311.1 - 2, 3 712
          BC141837.1 - 3, 4 364
          BC142673.1 - 3, 4 364
          BC144599.1 - 7, 8 6319 9053129
          BC144600.1 - 7, 8 6172 9053130
          BC150627.1 - 7, 8 6172 9056997
          CN420638.1 - 2, 3 560
          DR001720.1 - 2, 3 548

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          cell proliferation
          cerebral cortex development
          nuclear hormone receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0