Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BCL2L12 SIRNA Assays ›
          Gene Symbol:
          BCL2L12
          Gene Name:
          BCL2 like 12
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 49665072 - 49673916 on Build GRCh38
          UniGene ID:
          Hs.289052
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 36930

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BCL2L12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001040668.1 NP_001035758.1 5 1347
          NM_001282516.1 NP_001269445.1 5 1350
          NM_001282517.1 NP_001269446.1 4 1207
          NM_001282519.1 NP_001269448.1 5 1350
          NM_138639.1 NP_619580.1 5 1350
          NR_104200.1 4 1207
          NR_104201.1 5 1350
          NR_104202.1 4 1207
          NR_104203.1 4 1207
          XM_017027345.1 XP_016882834.1 5 1417
          XM_017027346.1 XP_016882835.1 4 1277
          GenBank mRNA BC007724.2 - 5 485
          BC104004.1 - 5 923 40024338
          BC104005.1 - 5 926 40024342
          BC104006.1 - 5 926 40024345
          BI159786.1 - 4 373
          FJ868798.1 - 5 668
          FJ868799.1 - 4 525
          FJ868800.1 - 5 668
          FJ868801.1 - 4 525
          FJ868802.1 - 5 668
          FJ868803.1 - 4 525
          KR820245.1 - 3 1504
          KR820246.1 - 3 1504
          KR820247.1 - 3 1504
          KR820248.1 - 3 1504
          KR820250.1 - 3 1553
          KR820251.1 - 3 1553
          KR820252.1 - 3 1553
          KR820253.1 - 3 1553
          KR820255.1 - 4 1285
          KR820256.1 - 4 1285
          KR820257.1 - 4 1285
          KR820259.1 - 4 1334
          KR820260.1 - 4 1334
          KR820261.1 - 4 1334
          KR820262.1 - 4 1334
          KR820264.1 - 4 1282
          KR820265.1 - 4 1282
          KR820267.1 - 4 1331
          KR820268.1 - 4 1331
          KR820269.1 - 4 1331
          KR820271.1 - 4 887
          KR820272.1 - 4 887
          KR820273.1 - 4 887
          KR820275.1 - 4 936
          KR820276.1 - 4 936
          KR820277.1 - 4 936
          KR820279.1 - 5 717
          KR820280.1 - 5 717
          KR820281.1 - 5 717
          KR820282.1 - 5 665
          KR820283.1 - 5 665
          KR820285.1 - 5 714
          KR820286.1 - 5 714
          KR820287.1 - 5 714
          KR820289.1 - 3 1142
          KR820290.1 - 3 1142
          KR820291.1 - 3 1142
          KR820292.1 - 3 1142

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          inhibition of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of cellular senescence
          p53 binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0