Search
Search

CANT1| Gene Symbol: CANT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159772.1 | NP_001153244.1 | 6 | 1685 | ||
| NM_001159773.1 | NP_001153245.1 | 5 | 1546 | |||
| NM_138793.3 | NP_620148.1 | 4 | 1741 | |||
| XM_005257021.1 | XP_005257078.1 | 5 | 1809 | |||
| XM_005257022.1 | XP_005257079.1 | 6 | 1664 | |||
| XM_006721683.1 | XP_006721746.1 | 7 | 1832 | |||
| XM_011524291.1 | XP_011522593.1 | 7 | 2137 | |||
| XM_011524293.1 | XP_011522595.1 | 6 | 2027 | |||
| XM_011524294.1 | XP_011522596.1 | 5 | 1502 | |||
| XM_011524295.2 | XP_011522597.1 | 7 | 1800 | |||
| GenBank mRNA | AF328554.2 | - | 6 | 1647 | ||
| AJ312208.1 | - | 3 | 826 | |||
| AK074687.1 | - | 4 | 1680 | |||
| AK295930.1 | - | 6 | 1369 | |||
| BC005104.1 | - | 1 | 180 | |||
| BC017655.1 | - | 5 | 1459 | 3898917 | ||
| BC065038.1 | - | 4 | 1362 | 6148580 | ||