Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VANGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172411.1 | NP_001165882.1 | 3 | 400 | ||
NM_001172412.1 | NP_001165883.1 | 3 | 344 | |||
NM_138959.2 | NP_620409.1 | 3 | 400 | |||
GenBank mRNA | AB057596.1 | - | 3 | 344 | ||
AB075805.1 | - | 2 | 141 | |||
AF481859.1 | - | 3 | 362 | |||
BC032773.1 | - | 1 | 68 | |||
BC065272.1 | - | 3 | 377 | 6165098 | ||
BM552933.1 | - | 419 |