siRNA Details
Gene Symbol: ADAMTS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139055.3 | NP_620686.1 | 1 | 161 | ||
GenBank mRNA | AJ315733.1 | - | 1 | 161 | ||
BC109114.1 | - | 1 | 161 | 40035454 |

Gene Symbol: ADAMTS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139055.3 | NP_620686.1 | 1 | 161 | ||
GenBank mRNA | AJ315733.1 | - | 1 | 161 | ||
BC109114.1 | - | 1 | 161 | 40035454 |