Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BIN1 SIRNA Assays ›
          Gene Symbol:
          BIN1
          Gene Name:
          bridging integrator 1
          Gene Aliases:
          AMPH2, AMPHL, SH3P9
          Chromosome Location:
          Chr.2: 127048023 - 127107400 on Build GRCh38
          UniGene ID:
          Hs.193163
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 37063

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BIN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001320632.1 NP_001307561.1 1, 2 470
          NM_001320633.1 NP_001307562.1 1, 2 470
          NM_001320640.1 NP_001307569.1 1, 2 470
          NM_001320641.1 NP_001307570.1 1, 2 470
          NM_004305.3 NP_004296.1 1, 2 470
          NM_139343.2 NP_647593.1 1, 2 470
          NM_139344.2 NP_647594.1 1, 2 470
          NM_139345.2 NP_647595.1 1, 2 470
          NM_139346.2 NP_647596.1 1, 2 470
          NM_139347.2 NP_647597.1 1, 2 470
          NM_139348.2 NP_647598.1 1, 2 470
          NM_139349.2 NP_647599.1 1, 2 470
          NM_139350.2 NP_647600.1 1, 2 470
          NM_139351.2 NP_647601.1 1, 2 470
          XM_005263642.3 XP_005263699.1 1, 2 543
          XM_005263643.3 XP_005263700.1 1, 2 543
          XM_006712425.3 XP_006712488.1 1, 2 543
          XM_006712426.3 XP_006712489.1 1, 2 543
          XM_006712428.3 XP_006712491.1 1, 2 543
          XM_006712429.3 XP_006712492.1 1, 2 543
          XM_006712431.3 XP_006712494.1 1, 2 543
          XM_006712432.3 XP_006712495.1 1, 2 543
          XM_006712433.3 XP_006712496.1 1, 2 543
          XM_006712434.3 XP_006712497.1 1, 2 543
          XM_017003820.1 XP_016859309.1 1, 2 543
          XM_017003821.1 XP_016859310.1 1, 2 543
          XM_017003822.1 XP_016859311.1 1, 2 543
          XM_017003827.1 XP_016859316.1 1, 2 543
          XM_017003828.1 XP_016859317.1 1, 2 543
          GenBank mRNA AF001383.1 - 1, 2 257
          AF004015.1 - 1, 2 86
          AF043898.1 - 1, 2 86
          AF043899.1 - 1, 2 86
          AF043900.1 - 1, 2 86
          AF043901.1 - 1, 2 86
          AF068914.1 - 1, 2 146
          AF068915.1 - 1, 2 146
          AF068916.1 - 1, 2 146
          AF068917.1 - 1, 2 146
          AF068918.1 - 1, 2 146
          AK295300.1 - 1, 2 188
          AL537027.3 - 1, 2 224
          AL713697.1 - 1, 2 431
          BC004101.2 - 1, 2 146 3688734
          BM742027.1 - 1, 2 188
          BT006865.1 - 1, 2 86
          CF594020.1 - 502
          U68485.1 - 1, 2 132
          U87558.1 - 1, 2 107

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          nucleus organization
          cell proliferation
          viral process
          regulation of endocytosis
          muscle cell differentiation
          positive regulation of apoptotic process
          regulation of neuron differentiation
          positive regulation of astrocyte differentiation
          lipid tube assembly
          regulation of cell cycle arrest
          protein binding
          identical protein binding
          tau protein binding
          actin filament binding
          RNA polymerase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0