Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BIN1 SIRNA Assays ›
          Gene Symbol:
          BIN1
          Gene Name:
          bridging integrator 1
          Gene Aliases:
          AMPH2, AMPHL, SH3P9
          Chromosome Location:
          Chr.2: 127048023 - 127107400 on Build GRCh38
          UniGene ID:
          Hs.193163
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 37153

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BIN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001320632.1 NP_001307561.1 2 497
          NM_001320633.1 NP_001307562.1 2 497
          NM_001320634.1 NP_001307563.1 2 263
          NM_001320640.1 NP_001307569.1 2 497
          NM_001320641.1 NP_001307570.1 2 497
          NM_001320642.1 NP_001307571.1 2 991
          NM_004305.3 NP_004296.1 2 497
          NM_139343.2 NP_647593.1 2 497
          NM_139344.2 NP_647594.1 2 497
          NM_139345.2 NP_647595.1 2 497
          NM_139346.2 NP_647596.1 2 497
          NM_139347.2 NP_647597.1 2 497
          NM_139348.2 NP_647598.1 2 497
          NM_139349.2 NP_647599.1 2 497
          NM_139350.2 NP_647600.1 2 497
          NM_139351.2 NP_647601.1 2 497
          XM_005263642.3 XP_005263699.1 2 570
          XM_005263643.3 XP_005263700.1 2 570
          XM_005263645.2 XP_005263702.1 2 54
          XM_006712425.3 XP_006712488.1 2 570
          XM_006712426.3 XP_006712489.1 2 570
          XM_006712427.2 XP_006712490.1 2 54
          XM_006712428.3 XP_006712491.1 2 570
          XM_006712429.3 XP_006712492.1 2 570
          XM_006712431.3 XP_006712494.1 2 570
          XM_006712432.3 XP_006712495.1 2 570
          XM_006712433.3 XP_006712496.1 2 570
          XM_006712434.3 XP_006712497.1 2 570
          XM_011510975.2 XP_011509277.1 2 1203
          XM_017003819.1 XP_016859308.1 2 384
          XM_017003820.1 XP_016859309.1 2 570
          XM_017003821.1 XP_016859310.1 2 570
          XM_017003822.1 XP_016859311.1 2 570
          XM_017003823.1 XP_016859312.1 2 54
          XM_017003824.1 XP_016859313.1 2 54
          XM_017003825.1 XP_016859314.1 2 384
          XM_017003826.1 XP_016859315.1 2 54
          XM_017003827.1 XP_016859316.1 2 570
          XM_017003828.1 XP_016859317.1 2 570
          GenBank mRNA AF001383.1 - 2 284
          AF004015.1 - 2 113
          AF043898.1 - 2 113
          AF043899.1 - 2 113
          AF043900.1 - 2 113
          AF043901.1 - 2 113
          AF068914.1 - 2 173
          AF068915.1 - 2 173
          AF068916.1 - 2 173
          AF068917.1 - 2 173
          AF068918.1 - 2 173
          AK295300.1 - 2 215
          AK301153.1 - 2 263
          AL537027.3 - 2 251
          AL713697.1 - 2 458
          BC004101.2 - 2 173 3688734
          BM742027.1 - 2 215
          BT006865.1 - 2 113
          CF594020.1 - 529
          U68485.1 - 2 159
          U87558.1 - 2 134

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          nucleus organization
          cell proliferation
          viral process
          regulation of endocytosis
          muscle cell differentiation
          positive regulation of apoptotic process
          regulation of neuron differentiation
          positive regulation of astrocyte differentiation
          lipid tube assembly
          regulation of cell cycle arrest
          protein binding
          identical protein binding
          tau protein binding
          actin filament binding
          RNA polymerase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nkcmf:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0