Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RGS3 SIRNA Assays ›
Gene Symbol:
RGS3
Gene Name:
regulator of G-protein signaling 3
Gene Aliases:
C2PA, RGP3
Chromosome Location:
Chr.9: 113444729 - 113597743 on Build GRCh38
UniGene ID:
Hs.494875
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
37235
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RGS3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001276260.1
NP_001263189.1
4
1500
NM_001276261.1
NP_001263190.1
12
1424
NM_001276262.1
NP_001263191.1
2
148
NM_001282922.1
NP_001269851.1
5
1358
NM_001282923.1
NP_001269852.1
20
2968
NM_001322215.1
NP_001309144.1
4
1465
NM_130795.3
NP_570613.2
13
2402
NM_134427.2
NP_602299.1
2
422
NM_144488.5
NP_652759.3
23
3306
NM_144489.3
NP_652760.2
2
434
XM_006717219.2
XP_006717282.1
22
3152
XM_006717220.1
XP_006717283.1
20
2928
XM_006717222.1
XP_006717285.1
13
2278
XM_006717225.1
XP_006717288.1
4
1282
XM_006717226.2
XP_006717289.1
5
1575
XM_006717230.3
XP_006717293.1
4
1164
XM_011518894.1
XP_011517196.1
5
1467
XM_011518896.1
XP_011517198.1
5
1829
XM_011518897.2
XP_011517199.1
6
1827
XM_011518898.1
XP_011517200.1
6
1551
XM_011518900.1
XP_011517202.1
5
1352
XM_011518901.1
XP_011517203.1
5
1383
XM_017015003.1
XP_016870492.1
22
3181
XM_017015004.1
XP_016870493.1
19
2676
XM_017015005.1
XP_016870494.1
7
1515
XM_017015006.1
XP_016870495.1
5
1731
GenBank mRNA
AF463495.1
-
13
2402
AF490838.1
-
2
434
AF490839.1
-
20
2780
AF490840.1
-
13
2252
AF493927.1
-
3
1058
AF493941.1
-
3
119
AK054584.1
-
2
411
AK096840.1
-
12
1344
AK123387.1
-
4
1473
AK125059.1
-
20
2939
AK128127.1
-
23
3304
AK226153.1
-
3
2872
AK289666.1
-
5
1194
AK309971.1
-
1
959
AY585192.1
-
3
1058
BC018072.1
-
2
225
BC019039.2
-
2
207
BC042555.1
-
13
2307
5175121
BE256545.1
-
2
148
BE298009.1
-
2
284
U27655.1
-
5
1345
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
inactivation of MAPK activity
regulation of G-protein coupled receptor protein signaling pathway
positive regulation of GTPase activity
GTPase activator activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-p89b7:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0