Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JSRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144616.3 | NP_653217.1 | 4 | 264 | ||
XM_017026285.1 | XP_016881774.1 | 4 | 658 | |||
XM_017026286.1 | XP_016881775.1 | 4 | 658 | |||
XM_017026287.1 | XP_016881776.1 | 4 | 240 | |||
GenBank mRNA | AK056978.1 | - | 4 | 258 | ||
BC021201.2 | - | 3 | 228 | 4106867 | ||
DA897543.1 | - | 4 | 265 | |||
DA898560.1 | - | 3 | 238 |