Search
Search

GNB2| Gene Symbol: GNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005273.3 | NP_005264.2 | 10 | 1652 | ||
| GenBank mRNA | AK056750.1 | - | 5 | 1774 | ||
| AK291592.1 | - | 10 | 1527 | |||
| BC010073.2 | - | 10 | 1576 | 4300639 | ||
| BC012348.1 | - | 10 | 1637 | 4556119 | ||
| BC068003.1 | - | 10 | 1562 | 6055863 | ||
| M16538.1 | - | 10 | 1522 | |||