Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144641.3 | NP_653242.3 | 3, 4 | 631 | ||
XM_005264879.2 | XP_005264936.1 | 3, 4 | 699 | |||
XM_005264881.1 | XP_005264938.1 | 3, 4 | 520 | |||
GenBank mRNA | AB474372.1 | - | 3, 4 | 599 | ||
AK056894.1 | - | 3, 4 | 520 | |||
AK096681.1 | - | 3, 4 | 526 | |||
AL832484.1 | - | 2, 3 | 442 | |||
DA869388.1 | - | 3, 4 | 521 |