Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BROX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288581.1 | NP_001275510.1 | 5 | 435 | ||
NM_144695.3 | NP_653296.2 | 5 | 729 | |||
XM_005273065.2 | XP_005273122.2 | 5 | 365 | |||
XM_005273069.4 | XP_005273126.1 | 4 | 601 | |||
XM_006711173.3 | XP_006711236.1 | 6 | 798 | |||
XM_011509213.2 | XP_011507515.1 | 4 | 673 | |||
XM_011509214.2 | XP_011507516.1 | 6 | 598 | |||
XM_017000374.1 | XP_016855863.1 | 7 | 923 | |||
XM_017000376.1 | XP_016855865.1 | 5 | 726 | |||
GenBank mRNA | AB276123.1 | - | 4 | 323 | ||
AK056983.1 | - | 5 | 718 | |||
AK304445.1 | - | 5 | 380 | |||
BC113635.1 | - | 5 | 466 | 8322687 | ||
BC113637.1 | - | 5 | 466 | 8322689 | ||
DC418391.1 | - | 5 | 435 |