Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB051812.1 | - | 24 | 4323 | ||
AB051813.1 | - | 25 | 4383 | |||
AB051814.1 | - | 25 | 4331 | |||
AB051815.1 | - | 24 | 4211 | |||
AF426259.1 | - | 4 | 1780 | |||
AF426260.1 | - | 5 | 1487 | |||
AL109788.1 | - | 1 | 1072 | |||
AY063456.1 | - | 6 | 1803 | |||
BC067102.1 | - | 24 | 4187 | 5925140 |