Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TACC2 SIRNA Assays ›
Gene Symbol:
TACC2
Gene Name:
transforming acidic coiled-coil containing protein 2
Gene Aliases:
AZU-1, ECTACC
Chromosome Location:
Chr.10: 121989174 - 122254545 on Build GRCh38
UniGene ID:
Hs.501252
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
3777
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TACC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291876.1
NP_001278805.1
8
6419
NM_001291877.1
NP_001278806.1
8
6566
NM_001291878.1
NP_001278807.1
3
927
NM_001291879.1
NP_001278808.1
5
777
NM_006997.3
NP_008928.1
3
927
NM_206860.2
NP_996742.1
3
927
NM_206861.2
NP_996743.1
6
992
NM_206862.3
NP_996744.3
9
6554
XM_005269388.1
XP_005269445.1
10
6378
XM_005269389.4
XP_005269446.1
10
6902
XM_005269390.1
XP_005269447.1
10
6378
XM_005269391.1
XP_005269448.1
9
6318
XM_005269392.1
XP_005269449.1
10
6378
XM_005269393.1
XP_005269450.1
10
6378
XM_005269394.1
XP_005269451.1
9
6252
XM_005269395.1
XP_005269452.1
9
6243
XM_005269396.1
XP_005269453.1
10
6378
XM_005269397.1
XP_005269454.1
8
6117
XM_005269399.1
XP_005269456.1
9
6243
XM_006717548.1
XP_006717611.1
10
6378
XM_006717552.1
XP_006717615.1
10
6378
XM_006717553.1
XP_006717616.1
10
6378
XM_006717554.1
XP_006717617.1
10
6378
XM_011539126.1
XP_011537428.1
10
6375
XM_011539128.1
XP_011537430.1
8
951
XM_011539129.1
XP_011537431.1
7
816
XM_011539130.1
XP_011537432.1
7
816
XM_011539131.1
XP_011537433.1
3
789
XM_011539132.1
XP_011537434.1
3
789
XM_011539133.1
XP_011537435.1
4
730
XM_011539134.1
XP_011537436.1
4
1001
XM_017015484.1
XP_016870973.1
10
6378
XM_017015485.1
XP_016870974.1
10
6378
XM_017015486.1
XP_016870975.1
10
6378
XM_017015487.1
XP_016870976.1
10
6378
XM_017015488.1
XP_016870977.1
8
6117
XM_017015489.1
XP_016870978.1
8
6117
XM_017015491.1
XP_016870980.1
3
929
XM_017015492.1
XP_016870981.1
3
929
XM_017015493.1
XP_016870982.1
3
789
XM_017015494.1
XP_016870983.1
3
929
XM_017015495.1
XP_016870984.1
3
789
XM_017015496.1
XP_016870985.1
4
730
XM_017015497.1
XP_016870986.1
4
1001
XM_017015498.1
XP_016870987.1
3
866
XM_017015499.1
XP_016870988.1
5
970
GenBank mRNA
AF095791.2
-
3
514
AF176646.1
-
4
915
AF220152.2
-
1
591
AF528098.1
-
6
973
AF528099.1
-
9
6535
AK295631.1
-
3
710
AL833304.1
-
8
5653
BC039311.1
-
3
927
BC141837.1
-
4
579
BC142673.1
-
4
579
BC144599.1
-
8
6534
9053129
BC144600.1
-
8
6387
9053130
BC150627.1
-
8
6387
9056997
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
cell proliferation
cerebral cortex development
nuclear hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0