Search
Search

FAM84A| Gene Symbol: FAM84A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145175.2 | NP_660158.2 | 2 | 572 | ||
| GenBank mRNA | AF370404.1 | - | 2 | 517 | ||
| AJ417080.1 | - | 2 | 526 | |||
| AK092654.1 | - | 2 | 541 | |||
| AK314463.1 | - | 2 | 541 | |||
| BC026346.1 | - | 1 | 566 | 4812838 | ||
| BC029933.1 | - | 2 | 526 | |||
| BC052284.1 | - | 2 | 521 | 5454643 | ||