Search
Search

C10orf35| Gene Symbol: C10orf35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145306.2 | NP_660349.1 | 3 | 257 | ||
| XM_005269606.1 | XP_005269663.1 | 3 | 541 | |||
| XM_005269608.3 | XP_005269665.1 | 2 | 195 | |||
| XM_011539455.2 | XP_011537757.1 | 3 | 798 | |||
| XM_017015836.1 | XP_016871325.1 | 4 | 429 | |||
| GenBank mRNA | BC013587.1 | - | 3 | 246 | 3896656 | |
| CK903062.1 | - | 2 | 196 | |||
| DB465414.1 | - | 3 | 256 | |||