Search
Search

GALNTL5| Gene Symbol: GALNTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145292.3 | NP_660335.2 | 8, 9 | 1454 | ||
| NR_033169.1 | 9, 10 | 1703 | ||||
| XM_006715862.3 | XP_006715925.1 | 9, 10 | 1529 | |||
| XM_011515849.2 | XP_011514151.1 | 7, 8 | 1141 | |||
| XM_011515852.2 | XP_011514154.1 | 7, 8 | 1277 | |||
| XM_011515853.2 | XP_011514155.1 | 6, 7 | 1035 | |||
| XM_017011793.1 | XP_016867282.1 | 8, 9 | 2728 | |||
| XM_017011794.1 | XP_016867283.1 | 7, 8 | 1026 | |||
| GenBank mRNA | AF440400.1 | - | 7, 8 | 1205 | ||
| AF440404.1 | - | 6, 7 | 1145 | |||
| AK093091.1 | - | 11, 12 | 1997 | |||
| AK315667.1 | - | 9, 10 | 1663 | |||
| BC022021.2 | - | 8, 9 | 1407 | 4825619 | ||