Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC30A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152275.3 | NP_689488.3 | 1 | 1945 | ||
GenBank mRNA | AK057530.1 | - | 2 | 1582 | ||
AK125104.1 | - | 2 | 1649 | |||
AK292395.1 | - | 1 | 1929 | |||
BC042848.1 | - | 1 | 1949 | 4831326 | ||
BC048006.1 | - | 1 | 1719 | 5299020 |