Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100873.1 | NP_001094343.1 | 3 | 435 | ||
NM_152337.2 | NP_689550.2 | 4 | 524 | |||
XM_005255792.2 | XP_005255849.1 | 4 | 472 | |||
XM_011522848.2 | XP_011521150.2 | 4 | 662 | |||
XM_011522849.2 | XP_011521151.2 | 4 | 691 | |||
XM_017022916.1 | XP_016878405.1 | 4 | 472 | |||
XM_017022917.1 | XP_016878406.1 | 3 | 422 | |||
GenBank mRNA | AK057264.1 | - | 4 | 456 | ||
AK295411.1 | - | 2 | 119 | |||
BC064143.1 | - | 3 | 381 | 30389327 | ||
DA525473.1 | - | 2 | 187 |