siRNA Details
Gene Symbol: SOX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006943.3 | NP_008874.2 | 1 | 3141 | ||
GenBank mRNA | AW296348.1 | - | 1 | 191 | ||
BC028103.1 | - | 2 | 1765 | |||
U35612.1 | - | 1 | 3117 |

More Information
Other Species Targeted

Panther Classification:
