Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152433.3 | NP_689646.2 | 2, 3 | 680 | ||
NM_198439.2 | NP_940841.1 | 3, 4 | 881 | |||
XM_006718767.3 | XP_006718830.1 | 3, 4 | 911 | |||
XM_011542618.2 | XP_011540920.1 | 2, 3 | 525 | |||
GenBank mRNA | AK092993.1 | - | 2, 3 | 283 | ||
AK289915.1 | - | 3, 4 | 491 | |||
BC062667.1 | - | 3, 4 | 573 | |||
BC107691.1 | - | 3, 4 | 492 | |||
BC117466.1 | - | 2, 3 | 379 | 40126017 | ||
BX640672.1 | - | 2, 3 | 401 | |||
BX647830.1 | - | 1, 2 | 46 | |||
CD104687.1 | - | 2, 3 | 352 |