Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC44A3 SIRNA Assays ›
Gene Symbol:
SLC44A3
Gene Name:
solute carrier family 44 member 3
Gene Aliases:
CTL3
Chromosome Location:
Chr.1: 94820342 - 94895247 on Build GRCh38
UniGene ID:
Hs.483423
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
38710
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC44A3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114106.2
NP_001107578.1
8
951
NM_001258340.1
NP_001245269.1
8
951
NM_001258341.1
NP_001245270.1
8
855
NM_001258342.1
NP_001245271.1
7
843
NM_001258343.1
NP_001245272.1
7
747
NM_001301079.1
NP_001288008.1
7
790
NM_152369.4
NP_689582.2
7
886
XM_005270440.2
XP_005270497.1
8
846
XM_005270441.2
XP_005270498.1
8
888
XM_005270442.1
XP_005270499.1
8
942
XM_011540632.2
XP_011538934.1
7
815
XM_011540633.2
XP_011538935.1
7
653
XM_011540635.2
XP_011538937.1
8
942
XM_011540636.2
XP_011538938.1
8
846
XM_017000262.1
XP_016855751.1
7
828
XM_017000263.1
XP_016855752.1
8
888
XM_017000264.1
XP_016855753.1
8
942
XM_017000265.1
XP_016855754.1
8
846
XM_017000266.1
XP_016855755.1
8
942
GenBank mRNA
AK300193.1
-
7
684
AK301288.1
-
7
692
AK302381.1
-
5
535
AK303906.1
-
7
822
AK313574.1
-
8
839
AK316123.1
-
9
796
AL530322.3
-
8
943
AL540829.3
-
7
843
AY358659.1
-
4
405
BC033858.1
-
8
914
BC053877.1
-
7
809
5760394
BX648225.1
-
7
815
Back To Top
More Information
Panther Classification:
Molecular Function -
transporter
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidylcholine biosynthetic process
choline transport
transmembrane transport
protein binding
choline transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-kz7s5:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0