Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM210A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098801.1 | NP_001092271.1 | 5 | 1160 | ||
NM_152352.3 | NP_689565.2 | 4 | 1020 | |||
GenBank mRNA | AK055618.1 | - | 5 | 1116 | ||
AK309506.1 | - | 2 | 407 | |||
AK309819.1 | - | 2 | 294 | |||
DA573992.1 | - | 2 | 408 |