Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152376.4 | NP_689589.1 | 2 | 1187 | ||
XM_005245742.3 | XP_005245799.1 | 2 | 1691 | |||
XM_011540699.2 | XP_011539001.1 | 2 | 1395 | |||
GenBank mRNA | AK058158.1 | - | 2 | 1172 | ||
BC036417.1 | - | 1 | 597 | |||
BC047428.1 | - | 2 | 1182 | 5295853 | ||
BX648631.1 | - | 2 | 1177 |