Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFBR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024847.2 | NP_001020018.1 | 3 | 708 | ||
NM_003242.5 | NP_003233.4 | 2 | 633 | |||
XM_011534043.2 | XP_011532345.1 | 3 | 481 | |||
XM_011534045.2 | XP_011532347.1 | 2 | 261 | |||
XM_017007106.1 | XP_016862595.1 | 2 | 1847 | |||
GenBank mRNA | AK300383.1 | - | 3 | 609 | ||
AK304404.1 | - | 3 | 377 | |||
AK314102.1 | - | 2 | 549 | |||
AL554950.3 | - | 2 | 645 | |||
BC040499.1 | - | 2 | 634 | |||
CB988859.1 | - | 3 | 636 | |||
D28131.1 | - | 3 | 266 | |||
KC792576.1 | - | 2 | 290 | |||
KC792577.1 | - | 2 | 290 | |||
KC792578.1 | - | 2 | 290 | |||
KC792579.1 | - | 2 | 290 | |||
KC792580.1 | - | 2 | 290 | |||
KU178359.1 | - | 2 | 251 | |||
KU178360.1 | - | 3 | 326 | |||
M85079.1 | - | 2 | 586 |