Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAS SIRNA Assays ›
Gene Symbol:
FAS
Gene Name:
Fas cell surface death receptor
Gene Aliases:
ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6
Chromosome Location:
Chr.10: 88968429 - 89017061 on Build GRCh38
UniGene ID:
Hs.244139
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
3905
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAS
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000043.5
NP_000034.1
2
425
NM_001320619.1
NP_001307548.1
2
425
NM_152871.3
NP_690610.1
2
425
NM_152872.3
NP_690611.1
2
425
NR_028033.3
2
425
NR_028034.3
2
425
NR_028035.3
2
425
NR_028036.3
2
425
NR_135313.1
2
425
NR_135314.1
2
416
NR_135315.1
2
416
XM_006717819.3
XP_006717882.1
3
5107
XM_011539764.2
XP_011538066.1
3
496
XM_011539765.2
XP_011538067.1
3
284
XM_011539766.2
XP_011538068.1
3
236
XM_011539767.2
XP_011538069.1
2
159
GenBank mRNA
AB209361.1
-
3
3742
AK290978.1
-
2
353
AK311164.1
-
2
273
AK311424.1
-
2
416
AK311499.1
-
2
273
AV715411.1
-
2
261
AY495076.2
-
2
301
BC012479.1
-
2
417
4514272
CR450307.1
-
2
107
DA040126.1
-
2
284
DA699562.1
-
2
425
EU481974.2
-
2
301
FJ200481.1
-
2
107
FJ373045.1
-
2
107
FJ373046.1
-
2
107
FM246458.1
-
2
107
FM246459.1
-
2
107
M67454.1
-
2
301
X63717.1
-
2
327
X83490.1
-
2
132
X83491.1
-
2
132
X83492.1
-
2
132
X83493.1
-
2
132
X89101.1
-
2
107
Z47993.1
-
2
107
Z47994.1
-
2
107
Z47995.1
-
2
107
Z66556.1
-
2
107
Z70519.1
-
2
107
Z70520.1
-
2
107
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle atresia
immunoglobulin production
renal system process
protein complex assembly
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
activation-induced cell death of T cells
inflammatory cell apoptotic process
inflammatory response
immune response
signal transduction
spermatogenesis
brain development
aging
circadian rhythm
extrinsic apoptotic signaling pathway via death domain receptors
response to toxic substance
gene expression
regulation of gene expression
B cell mediated immunity
telencephalon development
dendrite regeneration
positive regulation of protein homooligomerization
response to lipopolysaccharide
tumor necrosis factor-mediated signaling pathway
regulation of cell proliferation
response to drug
ovulation cycle
regulation of apoptotic process
chordate embryonic development
positive regulation of apoptotic process
negative regulation of apoptotic process
regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of MAPK cascade
response to peptide hormone
negative thymic T cell selection
regulation of lymphocyte differentiation
regulation of myeloid cell differentiation
response to cycloheximide
spleen development
negative regulation of B cell activation
protein homooligomerization
response to glucocorticoid
maternal process involved in female pregnancy
positive regulation of lymphocyte apoptotic process
cellular response to hydrogen peroxide
response to growth factor
cellular response to phenylalanine
cellular response to mechanical stimulus
cellular response to cobalt ion
cellular response to lithium ion
cellular response to glucose stimulus
cellular response to interleukin-1
cellular response to estrogen stimulus
cellular response to hyperoxia
cellular response to hypoxia
cellular response to hydrostatic pressure
motor neuron apoptotic process
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
extrinsic apoptotic signaling pathway in absence of ligand
hepatocyte apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
liver regeneration
necroptotic signaling pathway
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
response to fluoride
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
protease binding
signal transducer activity
receptor activity
tumor necrosis factor-activated receptor activity
protein binding
kinase binding
protein complex binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0