Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166691.1 | NP_001160163.1 | 5 | 722 | ||
NM_003318.4 | NP_003309.2 | 5 | 710 | |||
XM_011536099.2 | XP_011534401.1 | 5 | 684 | |||
XM_011536100.2 | XP_011534402.1 | 5 | 710 | |||
XM_017011242.1 | XP_016866731.1 | 6 | 1239 | |||
GenBank mRNA | AK095599.1 | - | 5 | 1182 | ||
AK292460.1 | - | 5 | 685 | |||
AK315696.1 | - | 5 | 627 | |||
AX775733.1 | - | 5 | 682 | |||
BC000633.2 | - | 5 | 673 | 3343925 | ||
BC032858.2 | - | 5 | 675 | 5270012 |