Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LMNTD1 SIRNA Assays ›
Gene Symbol:
LMNTD1
Gene Name:
lamin tail domain containing 1
Gene Aliases:
IFLTD1, PAS1C1
Chromosome Location:
Chr.12: 25476082 - 25648604 on Build GRCh38
UniGene ID:
Hs.44647
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
39271
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LMNTD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145727.2
NP_001139199.1
3
298
NM_001145728.2
NP_001139200.1
5
784
NM_001145729.1
NP_001139201.1
5
784
NM_001256266.1
NP_001243195.1
5
696
NM_152590.3
NP_689803.2
4
665
XM_011520575.1
XP_011518877.1
7
890
XM_011520576.2
XP_011518878.1
7
890
XM_011520577.2
XP_011518879.1
7
1300
XM_011520578.2
XP_011518880.1
7
913
XM_011520579.2
XP_011518881.1
7
890
XM_011520580.1
XP_011518882.1
7
890
XM_011520582.1
XP_011518884.1
7
902
XM_011520583.1
XP_011518885.1
6
771
XM_011520584.1
XP_011518886.1
4
578
XM_011520585.2
XP_011518887.1
4
501
XM_011520587.1
XP_011518889.1
4
274
XM_011520588.1
XP_011518890.1
3
196
XM_017018891.1
XP_016874380.1
6
722
XM_017018892.1
XP_016874381.1
6
792
XM_017018893.1
XP_016874382.1
5
601
XM_017018894.1
XP_016874383.1
7
890
XM_017018895.1
XP_016874384.1
8
949
GenBank mRNA
AK093323.1
-
4
626
AK296816.1
-
3
290
AK302292.1
-
5
784
AK310411.1
-
5
782
BC037957.1
-
5
665
5744078
BP359528.1
-
2
117
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell proliferation
structural molecule activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0