Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152609.2 | NP_689822.2 | 10 | 2238 | ||
XM_005273081.3 | XP_005273138.2 | 10 | 2023 | |||
XM_005273083.4 | XP_005273140.1 | 9 | 1941 | |||
XM_011544110.2 | XP_011542412.1 | 10 | 2089 | |||
XM_011544111.1 | XP_011542413.1 | 10 | 2260 | |||
XM_011544112.1 | XP_011542414.1 | 9 | 1807 | |||
XM_011544113.1 | XP_011542415.1 | 9 | 1932 | |||
XM_011544114.2 | XP_011542416.1 | 9 | 1647 | |||
XM_017000487.1 | XP_016855976.1 | 11 | 2278 | |||
XM_017000488.1 | XP_016855977.1 | 10 | 2021 | |||
XM_017000489.1 | XP_016855978.1 | 10 | 2088 | |||
GenBank mRNA | AK056563.1 | - | 10 | 2071 | ||
AK074142.1 | - | 10 | 2002 | |||
AK091739.1 | - | 2 | 784 | |||
AL834246.2 | - | 10 | 2072 | |||
BX647629.1 | - | 10 | 2023 |