Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200.3 | NP_001191.1 | 3 | 1219 | ||
XM_011529323.2 | XP_011527625.1 | 2 | 1264 | |||
GenBank mRNA | BC069214.1 | - | 3 | 1219 | ||
KC294426.1 | - | 2 | 434 | |||
M22489.1 | - | 3 | 757 |