Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RICTOR SIRNA Assays ›
          Gene Symbol:
          RICTOR
          Gene Name:
          RPTOR independent companion of MTOR complex 2
          Gene Aliases:
          AVO3, PIA, hAVO3
          Chromosome Location:
          Chr.5: 38937920 - 39074421 on Build GRCh38
          UniGene ID:
          Hs.407926
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 39500

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RICTOR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001285439.1 NP_001272368.1 3 186
          NM_001285440.1 NP_001272369.1 3 186
          NM_152756.4 NP_689969.2 3 186
          XM_006714463.3 XP_006714526.1 3 199
          XM_011514005.2 XP_011512307.1 3 199
          XM_017009311.1 XP_016864800.1 2 119
          XM_017009312.1 XP_016864801.1 2 119
          XM_017009313.1 XP_016864802.1 2 140
          XM_017009314.1 XP_016864803.1 3 177
          XM_017009315.1 XP_016864804.1 3 177
          GenBank mRNA AK128358.1 - 2 119
          AY515854.1 - 3 182
          BC029608.1 - 3 177
          BC137163.1 - 3 179 9021160
          BC137164.1 - 3 179 9021161
          BC144509.1 - 3 179 9053036
          BP245264.1 - 3 186
          CR749280.1 - 3 167

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of endothelial cell proliferation
          embryo development
          regulation of gene expression
          peptidyl-serine phosphorylation
          establishment of cell polarity
          positive regulation of actin filament polymerization
          establishment or maintenance of actin cytoskeleton polarity
          T cell costimulation
          actin cytoskeleton reorganization
          TOR signaling
          positive regulation of TOR signaling
          regulation of actin cytoskeleton organization
          regulation of peptidyl-serine phosphorylation
          regulation of phosphorylation
          positive regulation of catalytic activity
          regulation of GTPase activity
          phosphatidylinositol-mediated signaling
          regulation of inflammatory response
          positive regulation of peptidyl-tyrosine phosphorylation
          regulation of protein kinase B signaling
          positive regulation of protein kinase B signaling
          regulation of establishment of cell polarity
          protein binding
          enzyme activator activity
          protein kinase binding
          ribosome binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0