Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPERT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286341.1 | NP_001273270.1 | 2 | 521 | ||
NM_001286342.1 | NP_001273271.1 | 3 | 487 | |||
NM_152719.2 | NP_689932.1 | 3 | 602 | |||
XM_011534971.2 | XP_011533273.1 | 4 | 1957 | |||
GenBank mRNA | AK093129.1 | - | 3 | 595 | ||
AK292350.1 | - | 2 | 823 | |||
BC027609.2 | - | 2 | 421 | 4829821 | ||
HY016268.1 | - | 3 | 487 | |||
HY026582.1 | - | 2 | 405 |