Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152679.3 | NP_689892.1 | 3 | 1674 | ||
GenBank mRNA | BC012048.1 | - | 3 | 1532 | 3502817 | |
BC019066.2 | - | 3 | 1585 | 4897519 |