Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152744.3 | NP_689957.3 | 45 | 7724 | ||
XM_017011837.1 | XP_016867326.1 | 39 | 6565 | |||
GenBank mRNA | AK055987.1 | - | 6 | 1795 | ||
AK074077.1 | - | 12 | 2664 | |||
AK074083.1 | - | 30 | 5361 |