Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153252.4 | NP_694984.4 | 20 | 2509 | ||
XM_005262113.3 | XP_005262170.1 | 20 | 2511 | |||
XM_017029384.1 | XP_016884873.1 | 16 | 2039 | |||
XM_017029385.1 | XP_016884874.1 | 20 | 2511 | |||
GenBank mRNA | AK095887.1 | - | 3 | 382 | ||
AY497046.1 | - | 20 | 2509 | |||
AY497047.1 | - | 17 | 2441 | |||
AY497048.1 | - | 20 | 2532 | |||
AY497049.1 | - | 19 | 2502 | |||
AY497050.1 | - | 20 | 2482 | |||
AY497051.1 | - | 17 | 2069 | |||
AY497052.1 | - | 22 | 2668 | |||
AY497053.1 | - | 18 | 2257 | |||
AY497054.1 | - | 17 | 2158 | |||
AY497055.1 | - | 20 | 2443 | |||
AY497056.1 | - | 17 | 2414 | |||
AY497057.1 | - | 16 | 2313 | |||
AY497058.1 | - | 16 | 2152 | |||
AY497059.1 | - | 18 | 2523 | |||
AY497060.1 | - | 14 | 2090 | |||
BC111490.1 | - | 20 | 2459 |