Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCORL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166139.2 | NP_001159611.1 | 7 | 1474 | ||
XM_017007964.1 | XP_016863453.1 | 8 | 1350 | |||
GenBank mRNA | AK055258.1 | - | 1 | 362 | ||
BC062661.1 | - | 1 | 585 |